
di-Sodium hydrogen phosphate

Synonym(s):
Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
pK Value
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing