
Desogestrel

Synonym(s):
13-Ethyl-11-methylene-18,19-dinor-17α-4-pregnen-20-yn-17-ol, Desogestrel
Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique