
Cytidine 5′-triphosphate disodium solution

Synonym(s):
CTP-Na2
Empirical Formula (Hill Notation):
C9H16N3O14P3Na2
CAS Number:
Molecular Weight:
529.14
Products
Brand
Color
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing