
CPCCOEt

Synonym(s):
7-Hydroxyiminocyclopropan[b]chromen-1α-carboxylic acid ethyl ester
Empirical Formula (Hill Notation):
C13H13NO4
CAS Number:
Molecular Weight:
247.25
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing