
cis-(−)-Limonene oxide

Synonym(s):
(−)-cis-Limonene 1,2-epoxide, (−)-cis-p-Mentha-1,8-diene 1,2-epoxide, (1S,2R,4S)-4-Isopropenyl-1-methyl-1-cyclohexene 1,2-epoxide, (1S,4S)-1,2-Epoxy-p-menth-8-ene, (1S,4S,6R)-1-Methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]heptane
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Formula Weight
Physical Form
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing