Skip to Content
Merck
Chloroquine Related Compound D certified reference material, pharmaceutical secondary standard

Chloroquine Related Compound D

Chloroquine Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
7-Chloro-4-{[4-(ethylamino)-1-methylbutyl]amino}quinoline, Monodesethylchloroqu​ine
Empirical Formula (Hill Notation):
C16H22ClN3
CAS Number:
Molecular Weight:
291.82

Products

Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade