
CDP-Star® Chemiluminescent Substrate

Synonym(s):
Disodium 2-chloro-5-(4-methoxyspiro[1,2-dioxetane-3,2′-(5-chlorotricyclo[3.3.1.13.7]decan])-4-yl]-1-phenyl phosphate
Empirical Formula (Hill Notation):
C18H19Cl2O7Na2P
CAS Number:
Molecular Weight:
495.20
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing