
Azithromycin Identity

Synonym(s):
Azithromycin
Empirical Formula (Hill Notation):
C38H72N2O12
CAS Number:
Molecular Weight:
748.98
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing