
Atorvastatin Related Compound C

Synonym(s):
(βR,δR)-2,3-Bis(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt
Empirical Formula (Hill Notation):
C66H66CaF4N4O10
CAS Number:
Molecular Weight:
1191.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2