Skip to Content
Merck
Antimony foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%

Antimony

Antimony foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%
Synonym(s):
Antimony
Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element
Quality Segment
Showing 1-20 of 57
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3