
Anti-β-Tubulin−FITC antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-β-Tubulin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing