
Androsterone

Synonym(s):
3α-Hydroxy-5α-androstan-17-one, 5α-Androstan-3α-ol-17-one
Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing