
Acetone- 2,4-DNPH solution

Synonym(s):
Acetone-2,4-dinitrophenylhydrazone solution
Empirical Formula (Hill Notation):
C9H10N4O4
CAS Number:
Molecular Weight:
238.20
Products
Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique