Skip to Content
Merck
4-Oxo-TEMPO-1-15N 98 atom % 15N

4-Oxo-TEMPO-1-15N

4-Oxo-TEMPO-1-15N 98 atom % 15N
Synonym(s):
4-Oxo-2,2,6,6-tetramethylpiperidine-1-15N-1-oxyl, TEMPONE-1-15N
Empirical Formula (Hill Notation):
C9H1615NO2
Molecular Weight:
171.22
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (Ā°C)
Physical Form
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing