Skip to Content
Merck
4-Methylcyclohexanone 99%

4-Methylcyclohexanone

4-Methylcyclohexanone 99%
Synonym(s):
Tetrahydro-p-cresol, 4-Methylcyclohexanone
Linear Formula:
4-(CH3)C6H9(=O)
CAS Number:
Molecular Weight:
112.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing