Skip to Content
Merck
4-Methyl-2-pentanol 98%

4-Methyl-2-pentanol

4-Methyl-2-pentanol 98%
Synonym(s):
3-MIC, Isobutyl methyl carbinol, MAOH, MIBC, 4-Methyl-2-pentanol
Linear Formula:
(CH3)2CHCH2CH(OH)CH3
CAS Number:
Molecular Weight:
102.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Special Grade
Showing 1-3 of 3