
3-Nitrobenzoic acid

Synonym(s):
3-Nitrobenzoic acid
Linear Formula:
3-(NO2)C6H4COOH
CAS Number:
Molecular Weight:
167.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing