
3-Acetyl-11-keto-β-boswellic acid

Synonym(s):
AKBA, 3-O-Acetyl-11-keto-β-boswellic acid, 11-keto-β-Boswellic acid acetate, 3α-Acetoxy-11-oxo-12-ursen-24-oic acid
Empirical Formula (Hill Notation):
C32H48O5
CAS Number:
Molecular Weight:
512.72
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing