Skip to Content
Merck
3,5-Dinitrobenzyl chloride purum, ≥97.0% (AT)

3,5-Dinitrobenzyl chloride

3,5-Dinitrobenzyl chloride purum, ≥97.0% (AT)
Synonym(s):
α-Chloro-3,5-dinitrotoluene
Linear Formula:
(O2N)2C6H3CH2Cl
CAS Number:
Molecular Weight:
216.58
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing