Skip to Content
Merck
2-Nitro-4-(trifluoromethyl)benzoic acid 98%

2-Nitro-4-(trifluoromethyl)benzoic acid

2-Nitro-4-(trifluoromethyl)benzoic acid 98%
Synonym(s):
2-Nitro-α,α,α-trifluor-p-toluic acid
Linear Formula:
O2NC6H3(CF3)CO2H
CAS Number:
Molecular Weight:
235.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing