Skip to Content
Merck
2-Cyanoethyl N,N,N′,N′-tetraisopropylphosphorodiamidite

2-Cyanoethyl N,N,N′,N′-tetraisopropylphosphorodiamidite

2-Cyanoethyl N,N,N′,N′-tetraisopropylphosphorodiamidite
Synonym(s):
Bis(diisopropylamino)(2-cyanoethoxy)phosphine
Linear Formula:
{[(CH3)2CH]2N}2POCH2CH2CN
CAS Number:
Molecular Weight:
301.41

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Functional Group
Markush Class
Markush Group