Skip to Content
Merck
2-(Chloromethyl)anthraquinone 98%

2-(Chloromethyl)anthraquinone

2-(Chloromethyl)anthraquinone 98%
Empirical Formula (Hill Notation):
C15H9ClO2
CAS Number:
Molecular Weight:
256.68

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing