Skip to Content
Merck
2-Butoxyethyl acetate 99%

2-Butoxyethyl acetate

2-Butoxyethyl acetate 99%
Synonym(s):
1-Acetoxy-2-butoxyethane, BCA, Ethylene glycol monobutyl ether acetate, 2-Butoxyethyl acetate
Linear Formula:
CH3COOCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
160.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing