
2-Aminopyridine-4-boronic acid, pinacol ester

Empirical Formula (Hill Notation):
C11H17BN2O2
CAS Number:
Molecular Weight:
220.08
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing