
2-Amino-2-norbornanecarboxylic acid

Synonym(s):
2-Aminobicyclo[2.2.1]heptane-2-carboxylic acid
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing