![2-[(Acetoxymethyl)allyl]trimethylsilane 99%](/deepweb/assets/sigmaaldrich/product/structures/162/702/2145c2ca-c80b-47c6-abf6-d030858f0cc2/640/2145c2ca-c80b-47c6-abf6-d030858f0cc2.png)
2-[(Acetoxymethyl)allyl]trimethylsilane
![2-[(Acetoxymethyl)allyl]trimethylsilane 99%](/deepweb/assets/sigmaaldrich/product/structures/162/702/2145c2ca-c80b-47c6-abf6-d030858f0cc2/640/2145c2ca-c80b-47c6-abf6-d030858f0cc2.png)
Synonym(s):
2-(Trimethylsilylmethyl)-2-propen-1-yl acetate, 2-(Trimethylsilylmethyl)allyl acetate, 2-Acetoxymethyl-3-(trimethylsilyl)propene
Linear Formula:
CH3CO2CH2C(=CH2)CH2Si(CH3)3
CAS Number:
Molecular Weight:
186.32
Products
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing