
2,4-Diamino-6-phenyl-1,3,5-triazine

Synonym(s):
6-Phenyl-1,3,5-triazine-2,4-diamine, Benzoguanamine
Empirical Formula (Hill Notation):
C9H9N5
CAS Number:
Molecular Weight:
187.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing