Skip to Content
Merck
(1R,2R,5R)-(+)-2-Hydroxy-3-pinanone 99%

(1R,2R,5R)-(+)-2-Hydroxy-3-pinanone

(1R,2R,5R)-(+)-2-Hydroxy-3-pinanone 99%
Synonym(s):
(1R,2R,5R)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing