
1-Methylbenzimidazole

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing