1-Methyl-3-trifluoromethyl-1H-pyrazole-4-boronic acid pinacol ester
Empirical Formula (Hill Notation):
C11H16BF3N2O2
CAS Number:
Molecular Weight:
276.06
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2