Skip to Content
Merck
1H,1H,2H,2H-Perfluorodecyltrimethoxysilane ≥95.0%

1H,1H,2H,2H-Perfluorodecyltrimethoxysilane

1H,1H,2H,2H-Perfluorodecyltrimethoxysilane ≥95.0%
Empirical Formula (Hill Notation):
C13H13F17O3Si
CAS Number:
Molecular Weight:
568.30

Products

Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing