
1,3-Di-Boc-2-(trifluoromethylsulfonyl)guanidine

Synonym(s):
N,N′-Bis(tert-butoxycarboyl)-N′′-trifylguanidine
Linear Formula:
CF3SO2N=C[NHCO2C(CH3)3]2
CAS Number:
Molecular Weight:
391.36
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing