Skip to Content
Merck
All Photos(1)

Key Documents

MT51343203

Mettler-Toledo InLab® redox micro ORP electrode

Micro Redox Electrode with Platinum Ring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
39121436
NACRES:
NB.61

material

glass shaft

description

Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL
S7 connection
Without cable

manufacturer/tradename

Mettler-Toledo 51343203

temperature measuring range

0-100 °C

L × diam.

100 mm × 6 mm

General description

Mettler Toledo InLab® Redox Micro is A combination electrode for measuring the oxidation/reduction power of a solution. Consists of a platinum ring indicator electrode and a silver/silver chloride reference electrode.

The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.

The combination design avoids use of separate reference and indicating electrodes.
The Toledo InLab® Redox Micro electrode has the innovative Safelock closing ring which gives easy access for re-filling reference electrolyte and complete sealing when closed.

Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Combination design
  • Safelock closing ring
  • Detachable cable

Legal Information

ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service