GC Analysis of 3-Hydroxybutyric Acid Methyl Ester Enantiomers on Astec® CHIRALDEX™ B-DP
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 μm (78023AST)
oven
80 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: 3-hydroxybutyric acid methyl ester enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC