Merck
  • Plasma sample based analysis of gastric cancer progression using targeted metabolomics.
Plasma sample based analysis of gastric cancer progression using targeted metabolomics.
Scientific reports (2017-12-21)
Sergio Lario, Maria José Ramírez-Lázaro, Daniel Sanjuan-Herráez, Anna Brunet-Vega, Carles Pericay, Lourdes Gombau, Félix Junquera, Guillermo Quintás, Xavier Calvet
ABSTRACT
Gastric carcinogenesis is a multifactorial process described as a stepwise progression from non-active gastritis (NAG), chronic active gastritis (CAG), precursor lesions of gastric cancer (PLGC) and gastric adenocarcinoma. Gastric cancer (GC) 5-year survival rate is highly dependent upon stage of disease at diagnosis, which is based on endoscopy, biopsy and pathological examinations. Non-invasive GC biomarkers would facilitate its diagnosis at early stages leading to improved GC prognosis. We analyzed plasma samples collected from 80 patients diagnosed with NAG without H. pylori infection (NAG-), CAG with H. pylori infection (CAG+), PLGC and GC. A panel of 208 metabolites including acylcarnitines, amino acids and biogenic amines, sphingolipids, glycerophospholipids, hexoses, and tryptophan and phenylalanine metabolites were quantified using two complementary quantitative approaches: Biocrates AbsoluteIDQ®p180 kit and a LC-MS method designed for the analysis of 29 tryptophan pathway and phenylalanine metabolites. Significantly altered metabolic profiles were found in GC patients that allowing discrimination from NAG-, CAG+ and PLGC patients. Pathway analysis showed significantly altered tryptophan and nitrogen metabolic pathways (FDR P < 0.01). Three metabolites (histidine, tryprophan and phenylacetylglutamine) discriminated between non-GC and GC groups. These metabolic signatures open new possibilities to improve surveillance of PLGC patients using a minimally invasive blood analysis.
Sigma-Aldrich
6-Hydroxymelatonin
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
Indole-3-acetamide, 98%
Sigma-Aldrich
3-Hydroxy-DL-kynurenine
Supelco
Serotonin, analytical standard
Sigma-Aldrich
PhenylAc-Gln-OH, AldrichCPR