Merck
  • Hydroxyhomocitrulline Is a Collagen-Specific Carbamylation Mark that Affects Cross-link Formation.
Hydroxyhomocitrulline Is a Collagen-Specific Carbamylation Mark that Affects Cross-link Formation.
Cell chemical biology (2017-09-26)
Yuki Taga, Keisuke Tanaka, Chieko Hamada, Masashi Kusubata, Kiyoko Ogawa-Goto, Shunji Hattori
ABSTRACT
Carbamylation is a non-enzymatic post-translational modification that physiologically occurs during aging and is a risk factor for various diseases. The most common product of carbamylation is homocitrulline (HCit), where a lysine (Lys) amino group has reacted with urea-derived cyanate. HCit has recently been detected in collagen; however, given that 15%-90% of total Lys in collagen is hydroxylated, it is unclear how hydroxylation affects collagen carbamylation. Here, we identified a collagen-specific carbamylation product, hydroxyhomocitrulline (HHCit), and showed that high levels of HHCit are correlated with age in rat tissue collagen and in vivo carbamylation in mice, as well as with the decline of kidney function in the serum of dialysis patients. Proteomic analysis of the carbamylated collagens identified α2(I) Lys
Sigma-Aldrich
Sodium cyanate, 96%