Merck
  • Multiresidue analysis of 24 Water Framework Directive priority substances by on-line solid phase extraction-liquid chromatography tandem mass spectrometry in environmental waters.
Multiresidue analysis of 24 Water Framework Directive priority substances by on-line solid phase extraction-liquid chromatography tandem mass spectrometry in environmental waters.
Journal of chromatography. A (2017-03-21)
Adrià Rubirola, Mª Rosa Boleda, Mª Teresa Galceran
ABSTRACT
This paper reports the development of a fully multiresidue and automated on-line solid phase extraction (SPE) - liquid chromatography tandem mass spectrometry (LC-MS/MS) method for the determination of 24 priority substances (PS) belonging to different classes (pesticides, hormones or pharmaceuticals) included in the Directive 2013/39/UE and the recent Watch List (Decision 2015/495) in water samples (drinking water, surface water, and effluent wastewaters). LC-MS/MS conditions and on-line SPE parameters such as sorbent type, sample and wash volumes were optimized. The developed method is highly sensitive (limits of detection between 0.1 and 1.4ngL
Product Description
Supelco
Acetamiprid-d3, PESTANAL®, analytical standard
Sigma-Aldrich
Estrone-2,4,16,16-d4, 95 atom % D
Supelco
Imidacloprid-d4, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachlorophenol-13C6, 99 atom % 13C, 98% (CP)
Supelco
Isoproturon-d6, PESTANAL®, analytical standard
Supelco
Clothianidin-d3, PESTANAL®, analytical standard