Merck
  • Targeted disruption of hepcidin in the liver recapitulates the hemochromatotic phenotype.
Targeted disruption of hepcidin in the liver recapitulates the hemochromatotic phenotype.
Blood (2014-03-22)
Sara Zumerle, Jacques R R Mathieu, Stéphanie Delga, Mylène Heinis, Lydie Viatte, Sophie Vaulont, Carole Peyssonnaux
ABSTRACT
Hepcidin is a 25-amino-acid peptide demonstrated to be the iron regulatory hormone capable of blocking iron absorption from the duodenum and iron release from macrophages. Mutations affecting hepcidin regulators or the hepcidin gene itself cause hemochromatosis, a common genetic disorder. Hepcidin is produced mainly by the liver, but many cells and tissues express low levels of the hormone. To determine the contribution of these hepcidin-producing tissues in body iron homeostasis, we have developed a new mouse model in which the hepcidin gene can be conditionally inactivated. Here we compare a liver-specific knockout (KO) mouse model with total KO mice. We show that the liver-specific KO mice fully recapitulate the severe iron overload phenotype observed in the total KO mice, with increased plasma iron and massive parenchymal iron accumulation. This result demonstrates that the hepatocyte constitutes the predominant reservoir for systemic hepcidin and that the other tissues are unable to compensate.
Sigma-Aldrich
Anti-FTL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.