Merck
  • Use of olefin cross-metathesis to release azide-containing sugars from solid support.
Use of olefin cross-metathesis to release azide-containing sugars from solid support.
Organic letters (2003-11-25)
Takuya Kanemitsu, Peter H Seeberger
ABSTRACT
[reaction: see text] The octenediol linker used during automated oligosaccharide assembly is cleaved by olefin cross-metathesis. Until now, this linker could not be applied to sugars containing azides. A detailed study of the cleavage reaction served as basis for the development of a new protocol using the [(H(2)Imes)(3-Br-py)(2)(Cl)(2)Ru=CHPh] catalyst and 1-pentene. Efficient release of azide-protected carbohydrates from solid support can be readily achieved.
Sigma-Aldrich
1-Pentene, ≥98.5% (GC)
Sigma-Aldrich
1-Pentene, 98%
Supelco
1-Pentene, analytical standard
Sigma-Aldrich
Grubbs Catalyst® M300