Merck
  • Comparative analysis of Erk phosphorylation suggests a mixed strategy for measuring phospho-form distributions.
Comparative analysis of Erk phosphorylation suggests a mixed strategy for measuring phospho-form distributions.
Molecular systems biology (2011-04-14)
Sudhakaran Prabakaran, Robert A Everley, Isabelle Landrieu, Jean-Michel Wieruszeski, Guy Lippens, Hanno Steen, Jeremy Gunawardena
ABSTRACT
The functional impact of multisite protein phosphorylation can depend on both the numbers and the positions of phosphorylated sites-the global pattern of phosphorylation or 'phospho-form'-giving biological systems profound capabilities for dynamic information processing. A central problem in quantitative systems biology, therefore, is to measure the 'phospho-form distribution': the relative amount of each of the 2(n) phospho-forms of a protein with n-phosphorylation sites. We compared four potential methods-western blots with phospho-specific antibodies, peptide-based liquid chromatography (LC) and mass spectrometry (MS; pepMS), protein-based LC/MS (proMS) and nuclear magnetic resonance spectroscopy (NMR)-on differentially phosphorylated samples of the well-studied mitogen-activated protein kinase Erk2, with two phosphorylation sites. The MS methods were quantitatively consistent with each other and with NMR to within 10%, but western blots, while highly sensitive, showed significant discrepancies with MS. NMR also uncovered two additional phosphorylations, for which a combination of pepMS and proMS yielded an estimate of the 16-member phospho-form distribution. This combined MS strategy provides an optimal mixture of accuracy and coverage for quantifying distributions, but positional isomers remain a challenging problem.
Sigma-Aldrich
Anti-MAP Kinase, Monophosphorylated Tyrosine antibody ,Mouse monoclonal, clone ERK-PY193, purified from hybridoma cell culture
Sigma-Aldrich
3T3/A31 Cell Lysate
Sigma-Aldrich
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal, clone MAPK-YT, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP Kinase, Non-Phosphorylated ERK antibody, Mouse monoclonal, clone ERK-NP2, purified from hybridoma cell culture