Merck
  • Synthesis and Biological Evaluation of Fluorinated 3-Phenylcoumarin-7-O-Sulfamate Derivatives as Steroid Sulfatase Inhibitors.
Synthesis and Biological Evaluation of Fluorinated 3-Phenylcoumarin-7-O-Sulfamate Derivatives as Steroid Sulfatase Inhibitors.
Chemical biology & drug design (2015-08-19)
Sebastian Demkowicz, Mateusz Daśko, Witold Kozak, Katarzyna Krawczyk, Dariusz Witt, Maciej Masłyk, Konrad Kubiński, Janusz Rachon
ABSTRACT
In the present work, we report the initial results of our study on a series of 3-phenylcoumarin sulfamate-based compounds containing C-F bonds as novel inhibitors of steroid sulfatase. The new compounds are potent steroid sulfatase inhibitors, possessing more than 10 times higher inhibitory potency than coumarin-7-O-sulfamate. In the course of our investigation, compounds 2b and 2c demonstrated the highest inhibitory effect on the enzymatic steroid sulfatase assay; both had IC50 values of 0.27 μm (the IC50 value of coumarin-7-O-sulfamate is 3.5 μm, used as a reference).
Sigma-Aldrich
3,4-Difluorophenylacetic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(3,4,5-Trifluorophenyl)acetic acid, AldrichCPR