Merck
  • Natalizumab-associated progressive multifocal leukoencephalopathy in a patient with multiple sclerosis: a postmortem study.
Natalizumab-associated progressive multifocal leukoencephalopathy in a patient with multiple sclerosis: a postmortem study.
Journal of neuropathology and experimental neurology (2013-10-17)
Christian Wüthrich, Bogdan F Gh Popescu, Sarah Gheuens, Michael Marvi, Ronald Ziman, Stephen Pojen Denq, Mylyne Tham, Elizabeth Norton, Joseph E Parisi, Xin Dang, Claudia F Lucchinetti, Igor J Koralnik
ABSTRACT
Natalizumab, a monoclonal antibody directed against α4 integrins, has, to date, been associated with 399 cases of progressive multifocal leukoencephalopathy (PML) worldwide in patients receiving treatment for multiple sclerosis (MS). Because of the limited number of histologic studies, the possible interplay between MS and PML lesions has not been investigated. We report the clinical, radiologic, and histologic findings of an MS patient who developed PML after 32 months of natalizumab monotherapy. After withdrawal of natalizumab, she received plasma exchange, mefloquine, and mirtazapine but died soon thereafter. Postmortem examination was restricted to examination of the brain and spinal cord. Extensive PML lesions, characterized by the presence of JC virus DNA were found in the cerebral white matter and neocortex. Sharply demarcated areas of active PML lesions contained prominent inflammatory infiltrates composed of approximately equal numbers of CD4-positive and CD8-positive T cells, consistent with an immune reconstitution inflammatory syndrome. Conversely, all MS lesions identified were hypocellular, long-standing inactive plaques characterized by myelin loss, relative axonal preservation, and gliosis and, importantly, were devoid of JC virus DNA and active inflammation. Chronic inactive MS lesions were separate and distinct from nearby PML lesions. This case demonstrates the coexistence and apparent lack of interplay between chronic inactive MS and PML lesions, and that immune reconstitution inflammatory syndrome seems to affect the shape and appearance of PML but not MS lesions.
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Anti-CNP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit