Merck
  • Elevated MARCKS phosphorylation contributes to unresponsiveness of breast cancer to paclitaxel treatment.
Elevated MARCKS phosphorylation contributes to unresponsiveness of breast cancer to paclitaxel treatment.
Oncotarget (2015-05-28)
Ching-Hsien Chen, Chun-Ting Cheng, Yuan Yuan, Jing Zhai, Muhammad Arif, Lon Wolf R Fong, Reen Wu, David K Ann
ABSTRACT
Accumulating evidence has suggested that myristoylated alanine-rich C-kinase substrate (MARCKS) is critical for regulating multiple pathophysiological processes. However, the molecular mechanism underlying increased phosphorylation of MARCKS at Ser159/163 (phospho-MARCKS) and its functional consequence in neoplastic disease remain to be established. Herein, we investigated how phospho-MARCKS is regulated in breast carcinoma, and its role in the context of chemotherapy. In a screen of patients with breast tumors, we find that the abundance of phospho-MARCKS, not MARCKS protein per se, increased in breast cancers and positively correlated with tumor grade and metastatic status. Among chemotherapeutic agents, mitotic inhibitors, including paclitaxel, vincristine or eribulin, notably promoted phospho-MARCKS accumulation in multiple breast cancer cells. We further show that phospho-MARCKS acted upstream of Src activation upon paclitaxel exposure. Reduction of phospho-MARCKS by knockdown of MARCKS or pharmacological agents increased paclitaxel sensitivity. Particularly, a known phospho-MARCKS inhibitor, MANS peptide, was demonstrated to increase paclitaxel efficacy and attenuate angiogenesis/metastasis of xenografted breast cancer cells by decreasing abundance of phospho-MARCKS and messages of inflammatory mediators. Our data suggest that unresponsiveness of breast cancer to paclitaxel treatment is, at least in part, mediated by phospho-MARCKS and also provide an alternative therapeutic strategy against breast cancer by improving taxanes sensitivity.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Marcks
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human MARCKS
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%