Merck
  • Syntheses of imidazo-, oxa-, and thiazepine ring systems via ring-opening of aziridines/Cu-catalyzed C-N/C-C bond formation.
Syntheses of imidazo-, oxa-, and thiazepine ring systems via ring-opening of aziridines/Cu-catalyzed C-N/C-C bond formation.
The Journal of organic chemistry (2014-06-24)
Manas K Ghorai, Ashis Kumar Sahoo, Aditya Bhattacharyya
ABSTRACT
A simple and unpredicted synthetic pathway toward racemic and scalemic tetrahydrodibenzoimidazoazepines has been invented serendipitously proceeding through an S(N)2-type ring-opening of N-activated aziridines with 2-bromobenzylamine followed by a hitherto unprecedented cascade cyclization reaction sequence comprising a Cu-catalyzed cross dehydrogenation C-N coupling and an Ullmann C-C bond formation reaction. The tetrahydrobenzoxazepine and the tetrahydrobenzothiazepine derivatives have also been synthesized via the ring-opening of aziridines with 2-bromobenzyl alcohols and -mercaptan, respectively, followed by Cu-catalyzed N-arylation reaction.
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
2-Bromobenzyl alcohol, 99%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Copper(I) iodide, 98%
Copper(I) iodide ChemBeads