Merck
Subclinical atherosclerosis determinants in morbid obesity.
Nutrition, metabolism, and cardiovascular diseases : NMCD (2014-06-08)
I Megias-Rangil, J Merino, R Ferré, N Plana, M Heras, A Cabré, A Bonada, A Rabassa, L Masana
ABSTRACT
Obesity is associated with increased cardiovascular risk. However, the impact of morbid obesity on vascular structure and function is not well understood. This study was designed to appraise subclinical atherosclerosis markers, including carotid intima media thickness (cIMT), endothelial function, and arterial wall stiffness, and their determinants, in morbidly obese patients. In this cross-sectional study 194 overweight and obese patients were distributed in morbid-obese patients (MOP, n = 110), obese (OP, n = 84) and overweight patients (OwP, n = 33) groups. Demography, anthropometry, clinical and standard biochemical data were recorded. cIMT, endothelial function, defined as the small artery reactivity index (saRHI), and artery wall rigidity, studied by the augmentation index, were determined. More than 50% of the MOP, OP and OwP had a cIMT above the 75th percentile per age and gender. No differences in cIMT or saRHI were observed, although overweight and obese patients (OOP) had higher arterial rigidity compared with the morbid-obese patients. In a multivariate regression test, while cholesterol was the main determinant of cIMT in overweight and obese patients, glucose metabolism was the determinant in MOP. More than half of the population have a cIMT above general population ranges. OwP, OP and MOP have similar cIMT and saRHI. However, OOP have greater arterial wall rigidity. Dysglycemia is the main factor associated with subclinical atherosclerosis in MOP.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human E-Selectin ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Mouse E-Selectin ELISA Kit, for serum, plasma and cell culture supernatant