Merck
  • Multiple organ carcinogenicity of 1,3-butadiene in B6C3F1 mice after 60 weeks of inhalation exposure.
Multiple organ carcinogenicity of 1,3-butadiene in B6C3F1 mice after 60 weeks of inhalation exposure.
Science (New York, N.Y.) (1985-02-01)
J E Huff, R L Melnick, H A Solleveld, J K Haseman, M Powers, R A Miller
ABSTRACT
Groups of 50 male and 50 female B6C3F1 mice were exposed 6 hours per day, 5 days per week, for 60 to 61 weeks to air containing 0, 625, or 1250 parts per million 1,3-butadiene. These concentrations are somewhat below and slightly above the Occupational Safety and Health Administration standard of 1000 parts per million for butadiene. The study was designed for 104-week exposures but had to be ended early due to cancer-related mortality in both sexes at both exposure concentrations. There were early induction and significantly increased incidences of hemangiosarcomas of the heart, malignant lymphomas, alveolar-bronchiolar neoplasms, squamous cell neoplasms of the forestomach in males and females and acinar cell carcinomas of the mammary gland, granulosa cell neoplasms of the ovary, and hepatocellular neoplasms in females. Current workplace standards for exposure to butadiene should be reexamined in view of these findings.
Sigma-Aldrich
1,3-Butadiene, ≥99%
Pricing and availability is not currently available.