- Solid-phase reagents for selective monophosphorylation of carbohydrates and nucleosides.
Solid-phase reagents for selective monophosphorylation of carbohydrates and nucleosides.
The Journal of organic chemistry (2005-01-29)
Yousef Ahmadibeni, Keykavous Parang
PMID15675883
ABSTRACT
Two classes of aminomethyl polystyrene resin-bound linkers of p-acetoxybenzyl alcohol were subjected to reactions with 2-cyanoethyl N,N-diisopropylchlorophosphoramidite to produce the corresponding polymer-bound phosphitylating reagents. These were reacted with a number of unprotected nucleosides and carbohydrates in the presence of 1H-tetrazole. Oxidation with tert-butyl hydroperoxide followed by removal of the cyanoethoxy group with 1,8-diazabicyclo[5.4.0]undec-7-ene afforded the corresponding polymer-bound phosphate diesters. Acidic cleavage of the p-acetoxybenzyl alcohol linker yielded monophosphorylated products with high regioselectivity and trapped linkers on the resins that can be reused.