- Distribution and persistence of aminocarb in stream water, sediment and fish after application of three Matacil formulations.
Distribution and persistence of aminocarb in stream water, sediment and fish after application of three Matacil formulations.
Three aminocarb formulations, water-based Matacil 180F, oil-based Matacil 180F and oil-based Matacil 180D, were applied separately to the surface of a stream at 2.40 g a.i./min for 5 minutes. The highest aminocarb concentration in the top 1 cm of water was found 5 min after application of oil-based 180F (5000 ppb). The next highest concentration was 1862 ppb found in the top 1 cm layer of water 3 min after application of oil-based 180D, followed by 1306 ppb found 5 min after application of water-based 180F. Residue accumulations were first observed in sediment at 3 min and in fish tissues 0.5 h after application. The levels of accumulation were in the order of water-based 180F greater than oil-based 180F greater than oil-based 180-D. After the application of water-based 180F, the highest concentrations were 20.2 ppb in sediment after 5 min and 127 ppb in fish tissues after 3 h. Residues disappeared rapidly from all three matrices and could not be detected after a period of 3-25 h.