Merck
  • Molecular interaction of PCB153 to human serum albumin: insights from spectroscopic and molecular modeling studies.
Molecular interaction of PCB153 to human serum albumin: insights from spectroscopic and molecular modeling studies.
Journal of hazardous materials (2013-02-19)
Chao Han, Senbiao Fang, Huiming Cao, Yan Lu, Yaqiong Ma, Dongfeng Wei, Xiaoyun Xie, Xiaohua Liu, Xin Li, Dongqing Fei, Chunyan Zhao
ABSTRACT
Polychlorinated biphenyls (PCBs) possessed much potential hazard to environment because of its chemical stability and biological toxicity. Here, we identified the binding mode of a representative compound, PCB153, to human serum albumin (HSA) using fluorescence and molecular dynamics simulation methods. The fluorescence study showed that the intrinsic fluorescence of HSA was quenched by addition of PCB153 through a static quenching mechanism. The thermodynamic analysis proved the binding behavior was mainly governed by hydrophobic force. Furthermore, as evidenced by site marker displacement experiments using two probe compounds, it revealed that PCB153 acted exactly on subdomain IIIA (site II) of HSA. On the other hand, the molecular dynamics studies as well as free energy calculations made another important contribution to understand the conformational changes of HSA and the stability of HSA-PCB153 system. Molecular docking revealed PCB153 can bind in a large hydrophobic activity of subdomain IIIA by the hydrophobic interaction and hydrogen bond interactions between chlorine atoms and residue ASN391. The present work provided reasonable models helping us further understand the transporting, distribution and toxicity effect of PCBs when it spread into human blood serum.
Product Description
Supelco
PCB No 153, analytical standard
2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153), BCR®, certified reference material