Merck
  • Heterogeneous cationic liposomes modified with 3β-{N-[(N',N'-dimethylamino)ethyl]carbamoyl}cholesterol can induce partial conformational changes in messenger RNA and regulate translation in an Escherichia coli cell-free translation system.
Heterogeneous cationic liposomes modified with 3β-{N-[(N',N'-dimethylamino)ethyl]carbamoyl}cholesterol can induce partial conformational changes in messenger RNA and regulate translation in an Escherichia coli cell-free translation system.
Langmuir : the ACS journal of surfaces and colloids (2013-01-18)
Keishi Suga, Tomoyuki Tanabe, Hiroshi Umakoshi
ABSTRACT
The effect of cationic liposomes (CLs) on messenger RNA(mRNA) conformation and translation was studied, focusing on membrane heterogeneity. CLs, composed of 1,2-dioleoyl-sn-glycerol-3-phosphocholine/1,2-dioleoyl-3-timethylammonium propane (DOPC/DOTAP) and DOPC/3β-{N-[(N',N'-dimethylamino)ethyl]carbamoyl}cholesterol (DOPC/DC-Ch), inhibited mRNA translation in an Escherichia coli cell-free translation system. Analysis of the membrane fluidity and polarity indicated a heterogeneous DOPC/DC-Ch (70/30) membrane, while other CLs exhibited homogeneous disordered membranes. mRNA adsorbed onto DOPC/DC-Ch liposomes showed translational activity, while DOPC/DOTAP liposomes inhibited mRNA translation in proportion to its adsorption onto membranes. Dehydration of DOPC/DOTAP (70/30) and DOPC/DC-Ch (70/30) was observed in the presence of mRNA but not in the case of zwitterionic DOPC liposomes, indicating that mRNA binds in regions between the phosphate [-PO(2)(-)-] and carbonyl [-C=O-] moieties of lipids. UV resonance Raman spectroscopy suggests that adenine, cytosine, and guanine interact with DOPC/DOTAP (70/30) and DOPC/DC-Ch (70/30) but not with DOPC. Circular dichroism indicates that DOPC/DOTAP (70/30) extensively denatured the mRNA. In contrast, heterogeneous DOPC/DC-Ch (70/30) induced partial conformational changes but maintained the translational activity of mRNA.
Sigma-Aldrich
1,6-Diphenyl-1,3,5-hexatriene, 98%
Sigma-Aldrich
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Cholesteryl N-(2-dimethylaminoethyl)carbamate, ≥98% (TLC)
Supelco
2-Naphthylamine, analytical standard